3-Fluoro-2-methoxybenzyl bromide 97%, JRD

Danger

H32562-MD H32562-03
AAH32562-MDEA 87.78 USD
AAH32562-MD AAH32562-03
3-Fluoro-2-methoxybenzyl bromide 97%, JRD
3-Fluoro-2-methoxybenzyl bromide
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD09025370
CAS Number: 916420-50-3
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR