2,3-Dimethyl-4-fluoronitrobenzene 98%

Danger

H32557-03 H32557-MD H32557-06
AAH32557-MDEA 57.88 USD
AAH32557-MD AAH32557-06 AAH32557-03
2,3-Dimethyl-4-fluoronitrobenzene 98%
2,3-Dimethyl-4-fluoronitrobenzene
Formula: C₈H₈FNO₂
MW: 169.16 g/mol
Melting Pt: 39…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD05663712
CAS Number: 1736-87-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR