2,6-Dichloro-3-(trifluoromethyl)aniline 97%, JRD
|
Danger
|
H32552-03
H32552-MD
AAH32552-MDEA
68.98
USD
AAH32552-MD
AAH32552-03
2,6-Dichloro-3-(trifluoromethyl)aniline 97%, JRD
2,6-Dichloro-3-(trifluoromethyl)aniline
Formula:
C₇H₄Cl₂F₃N Storage Temperature: Ambient |
MDL Number:
MFCD11226279 CAS Number: 6656-72-0 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...