3'-Chloro-4'-ethoxy-5'-fluoroacetophenone 97%, JRD

Warning

H32546-06 H32546-03
AAH32546-03EA 28.49 USD
AAH32546-03 AAH32546-06
3'-Chloro-4'-ethoxy-5'-fluoroacetophenone 97%, JRD
3'-Chloro-4'-ethoxy-5'-fluoroacetophenone
Formula: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD09832347
CAS Number: 1017779-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR