4-Methyl-2-(trifluoromethyl)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H32534-03
AAH32534-03EA 149.99 USD
AAH32534-03
4-Methyl-2-(trifluoromethyl)benzoic acid 98%
4-Methyl-2-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631591
CAS Number: 120985-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR