2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: 3-Fluoro-5-(trifluoromethyl)pyridin-2-amine

H32532-MD H32532-06
AAH32532-06EA 556 USD
AAH32532-06 AAH32532-MD
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Formula: C₆H₄F₄N₂
MW: 180.10 g/mol
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD10699117
CAS Number: 852062-17-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR