Tributylborane 1 M in THF

Danger

H32518-AP
AAH32518-APEA 481.07 USD
AAH32518-AP
Tributylborane 1 M in THF
Tributylborane
Formula: C₁₂H₂₇B
Density: 0.834
Storage Temperature: Ambient
MDL Number: MFCD00009423
CAS Number: 122-56-5
UN: 3392
ADR: "4.2",I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR