2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid 98%

Warning

Synonyms: 2-tert-Butyl-4-methylthiazole-5-carboxylic acid

H32501-03 H32501-MD
AAH32501-MDEA 41.76 USD
AAH32501-MD AAH32501-03
2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid 98%
2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid
Formula: C₉H₁₃NO₂S
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD11207787
CAS Number: 427896-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR