4-Amino-3,5-difluorophenol 98%

Warning

H32493-03 H32493-MD
AAH32493-03EA 240.07 USD
AAH32493-03 AAH32493-MD
4-Amino-3,5-difluorophenol 98%
4-Amino-3,5-difluorophenol
Formula: C₆H₅F₂NO
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD03094491
CAS Number: 135086-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR