Agarose 6% B, beads

H32491-AU
AAH32491-AUEA 612.17 USD
AAH32491-AU
Agarose 6% B, beads
Agarose
Boiling Pt: 100 °C
Density: 1.040
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR