1H-Indazole-7-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals

Warning

H32463-MD H32463-03
AAH32463-03EA 468.84 USD
AAH32463-03 AAH32463-MD
1H-Indazole-7-carboxylic acid 95%
1H-Indazole-7-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06804572
CAS Number: 677304-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR