4-Ethoxy-2,3-difluorobenzaldehyde 97%, JRD

Supplier: Alfa Aesar
Warning

H32450-06 H32450-03
AAH32450-06EA 125 USD
AAH32450-06 AAH32450-03
4-Ethoxy-2,3-difluorobenzaldehyde 97%, JRD
4-Ethoxy-2,3-difluorobenzaldehyde
Formula: C₉H₈F₂O₂
MW: 186,16
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD09258700
CAS Number: 126162-95-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR