3-Chloro-4-(trifluoromethoxy)phenylacetic acid 97%, JRD

Supplier: Alfa Aesar
Warning

H32437-MD H32437-03
AAH32437-MDEA 85.12 USD
AAH32437-MD AAH32437-03
3-Chloro-4-(trifluoromethoxy)phenylacetic acid 97%, JRD
3-Chloro-4-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₆ClF₃O₃
MW: 254,59
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09832392
CAS Number: 1017779-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR