3-Bromo-5-(trifluoromethoxy)phenylacetic acid 96%, JRD

Warning

H32419-MD H32419-03
AAH32419-03EA 245.31 USD
AAH32419-03 AAH32419-MD
3-Bromo-5-(trifluoromethoxy)phenylacetic acid 96%, JRD
3-Bromo-5-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₆BrF₃O₃
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD11519378
CAS Number: 1092461-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR