2-Methyl-3-(trifluoromethyl)phenylacetonitrile 97%, JRD

Warning

H32412-MD H32412-03
AAH32412-MDEA 70.64 USD
AAH32412-MD AAH32412-03
2-Methyl-3-(trifluoromethyl)phenylacetonitrile 97%, JRD
2-Methyl-3-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃N
Melting Pt: 51…55 °C
Storage Temperature: Ambient
MDL Number: MFCD09832278
CAS Number: 1000515-00-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR