3-(4-(Difluoromethoxy)phenyl)propionic acid 97%, JRD

Warning

H32381-MD H32381-03
AAH32381-MDEA 81.03 USD
AAH32381-MD AAH32381-03
3-(4-(Difluoromethoxy)phenyl)propionic acid 97%, JRD
3-(4-(Difluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₁₀F₂O₃
MW: 216.18
Melting Pt: 66…69°C
Storage Temperature: Ambient
MDL Number: MFCD09863355
CAS Number: 1000932-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR