3-Chloro-5-(trifluoromethyl)pyridine 97%

Warning

H32376-06 H32376-03
AAH32376-03EA 49.73 USD
AAH32376-03 AAH32376-06
3-Chloro-5-(trifluoromethyl)pyridine 97%
3-Chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.55 g/mol
Boiling Pt: 50…51 °C/4 mm
Storage Temperature: Ambient
MDL Number: MFCD00042227
CAS Number: 85148-26-1
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR