3-Chloro-4-(trifluoromethyl)benzoyl chloride 97%, JRD
|
Danger
|
H32343-06
AAH32343-06EA
346.32
USD
AAH32343-06
3-Chloro-4-(trifluoromethyl)benzoyl chloride 97%, JRD
3-Chloro-4-(trifluoromethyl)benzoyl chloride
Formula:
C₈H₃Cl₂F₃O MW: 243.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519325 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...