2,3,5-Trifluorobenzenesulfonyl chloride 97%, JRD

Danger

Synonyms: 2,3,5-Trifluorobenzene-1-sulphonyl chloride2,3,5-Trifluorobenzene-1-sulfonyl chloride

H32331-MD H32331-03
AAH32331-03EA 247.53 USD
AAH32331-03 AAH32331-MD
2,3,5-Trifluorobenzenesulfonyl chloride 97%, JRD
2,3,5-Trifluorobenzenesulphonyl chloride
Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Melting Pt: 40…49 °C
Storage Temperature: Ambient
MDL Number: MFCD08532462
CAS Number: 914636-99-0
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR