(3,6-Dichloro-2-fluorophenyl)acetic acid 97%, JRD

Warning

H32318-MD H32318-03
AAH32318-03EA 200.91 USD
AAH32318-03 AAH32318-MD
(3,6-Dichloro-2-fluorophenyl)acetic acid 97%, JRD
(3,6-Dichloro-2-fluorophenyl)acetic acid
Formula: C₈H₅Cl₂FO₂
MW: 223.03
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD09025391
CAS Number: 916420-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR