Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Ethylamine 2.0 M in THF

Supplier: Alfa Aesar
Danger

H32306-AE H32306-AP
AAH32306-AEEA 80.9 USD InStock
AAH32306-AE AAH32306-AP
Ethylamine 2.0 M in THF
Ethylamine


Formula: C₂H₇N
MW: 45,08 g/mol
Boiling Pt: 17 °C (1013 hPa)
Melting Pt: -81.1 °C
Density: 0,856
Flash Pt: -34°C (-29°F)
Storage Temperature: Ambient
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7
UN: 2056
ADR: 3,II
Merck Index: 13,03797
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty