3-Fluoro-2-methoxybenzylamine 97%, JRD

Danger

H32293-MD H32293-03
AAH32293-MDEA 103.56 USD
AAH32293-MD AAH32293-03
3-Fluoro-2-methoxybenzylamine 97%, JRD
3-Fluoro-2-methoxybenzylamine
Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD09025368
CAS Number: 916420-49-0
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR