4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride 97%, JRD

Danger

H32280-03 H32280-MD
AAH32280-MDEA 58.97 USD
AAH32280-MD AAH32280-03
4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride 97%, JRD
4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride
Formula: C₈H₆ClF₃O₃S
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD11519323
CAS Number: 775288-85-2
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR