4-Amino-3-iodobenzoic acid 98%

Warning

H32263-03 H32263-06
AAH32263-03EA 126.2 USD
AAH32263-03 AAH32263-06
4-Amino-3-iodobenzoic acid 98%
4-Amino-3-iodobenzoic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01660550
CAS Number: 2122-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR