4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride 97%, JRD

Danger

H32242-MD H32242-03
AAH32242-03EA 174.27 USD
AAH32242-03 AAH32242-MD
4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride 97%, JRD
4-Methoxy-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₆ClF₃O₃S
MW: 274.64
Melting Pt: 52…55 °C
Storage Temperature: Ambient
MDL Number: MFCD07774167
CAS Number: 694514-21-1
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR