(4-Methoxy-3,5-dimethylphenyl)amine 98%

Danger

Synonyms: 3,5-Dimethyl-4-methoxyaniline4-Methoxy-3,5-dimethylaniline

H32232-MD H32232-03
AAH32232-03EA 245.12 USD
AAH32232-03 AAH32232-MD
(4-Methoxy-3,5-dimethylphenyl)amine 98%
(4-Methoxy-3,5-dimethylphenyl)amine
Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01907075
CAS Number: 39785-37-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR