5-Fluoro-2-(trifluoromethoxy)benzyl bromide 97%, JRD

Danger

H32201-MD H32201-03
AAH32201-03EA 165.39 USD
AAH32201-03 AAH32201-MD
5-Fluoro-2-(trifluoromethoxy)benzyl bromide 97%, JRD
5-Fluoro-2-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD11519337
CAS Number: 1092460-88-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR