3-Bromo-6-fluorochromone 97%

H32163-06 H32163-03
AAH32163-03EA 83.97 USD
AAH32163-03 AAH32163-06
3-Bromo-6-fluorochromone 97%
3-Bromo-6-fluorochromone
Formula: C₉H₄BrFO₂
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD03094003
CAS Number: 179111-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR