3'-Bromoacetophenone diethyl ketal 96% stabilized

Warning


Stabilization: Stabilised with potassium carbonate

H32157-14 H32157-06
AAH32157-06EA 79.35 USD
AAH32157-06 AAH32157-14
3'-Bromoacetophenone diethyl ketal 96% stabilized
3'-Bromoacetophenone diethyl ketal
Formula: C₁₂H₁₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD14636457
CAS Number: 480439-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR