4-(6-Chloro-4-pyrimidinyl)morpholine 98%

Warning

Synonyms: 4-(6-Chloropyrimidin-4-yl)morpholine

H32140-ME
AAH32140-MEEA 130.17 USD
AAH32140-ME
4-(6-Chloro-4-pyrimidinyl)morpholine 98%
4-(6-Chloro-4-pyrimidinyl)morpholine
Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00466219
CAS Number: 22177-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR