4-Bromo-5-nonanone 98%

Danger

H32136-09 H32136-03
AAH32136-09EA 411.81 USD
AAH32136-09 AAH32136-03
4-Bromo-5-nonanone 98%
4-Bromo-5-nonanone
Formula: C₉H₁₇BrO
Boiling Pt: 62 °C/1 mm
Density: 1.174
Flash Pt: 92 °C(198 °F)
Storage Temperature: Ambient
MDL Number: MFCD09757527
CAS Number: 42330-11-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR