2-Bromo-N-(4-chlorobenzyl)acetamide 95%

Warning

H32130-03 H32130-MD
AAH32130-MDEA 42.96 USD
AAH32130-MD AAH32130-03
2-Bromo-N-(4-chlorobenzyl)acetamide 95%
2-Bromo-N-(4-chlorobenzyl)acetamide
Formula: C₉H₉BrClNO
MW: 262.54 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD11226535
CAS Number: 934185-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR