6-Bromoanthranilic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-6-bromobenzoic acid

H32106-06 H32106-03
AAH32106-03EA 139.99 USD
AAH32106-03 AAH32106-06
6-Bromoanthranilic acid ≥98%
6-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618455
CAS Number: 20776-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR