3-(3-Fluoro-4-methylphenyl)propionic acid 97%, JRD

Warning

H32105-MD H32105-03
AAH32105-MDEA 64.74 USD
AAH32105-MD AAH32105-03
3-(3-Fluoro-4-methylphenyl)propionic acid 97%, JRD
3-(3-Fluoro-4-methylphenyl)propionic acid
Formula: C₁₀H₁₁FO₂
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09025364
CAS Number: 881189-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR