2-Fluoro-5-(trifluoromethoxy)aniline 97%, JRD
|
Danger
|
H32103-06
H32103-03
AAH32103-03EA
146.18
USD
AAH32103-03
AAH32103-06
2-Fluoro-5-(trifluoromethoxy)aniline 97%, JRD
2-Fluoro-5-(trifluoromethoxy)aniline
Formula:
C₇H₅F₄NO MW: 195.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832254 CAS Number: 116369-23-4 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...