Ethyl-1H-indazole-3-carboxylate 95%

Synonyms: Ethyl 1H-indazole-3-carboxylate

H32098-03 H32098-MD
AAH32098-MDEA 156.51 USD
AAH32098-MD AAH32098-03
Ethyl-1H-indazole-3-carboxylate 95%
Ethyl 1H-indazole-3-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD01138134
CAS Number: 4498-68-4
EINECS: 224-795-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR