5-Methyl-2-(trifluoromethyl)benzylamine 97%, JRD

Danger

H32063-MD H32063-03
AAH32063-03EA 179.82 USD
AAH32063-03 AAH32063-MD
5-Methyl-2-(trifluoromethyl)benzylamine 97%, JRD
5-Methyl-2-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃N
Storage Temperature: Ambient
MDL Number: MFCD09832358
CAS Number: 1017779-30-4
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR