2,4-Difluoro-3-methylbenzylamine 95%

Danger

H32060-MD H32060-06 H32060-03
AAH32060-MDEA 63.46 USD
AAH32060-MD AAH32060-06 AAH32060-03
2,4-Difluoro-3-methylbenzylamine 95%
2,4-Difluoro-3-methylbenzylamine
Formula: C₈H₉F₂N
Boiling Pt: 42/1 mm
Storage Temperature: Ambient
MDL Number: MFCD08275293
CAS Number: 847502-90-3
EINECS: 642-729-2
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR