2-Acetamido-6-methylpyridine 98%

Warning

Synonyms: N-(6-Methylpyridin-2-yl)acetamide

H32034-09 H32034-18
AAH32034-18EA 288.5 USD
AAH32034-18 AAH32034-09
2-Acetamido-6-methylpyridine 98%
2-Acetamido-6-methylpyridine
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00091881
CAS Number: 5327-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR