2-Fluoro-5-iodobenzaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

H32033-06 H32033-14 H32033-03
AAH32033-06EA 127.99 USD
AAH32033-06 AAH32033-14 AAH32033-03
2-Fluoro-5-iodobenzaldehyde 97+%
2-Fluoro-5-iodobenzaldehyde
Formula: C₇H₄FIO
MW: 250.01 g/mol
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03093999
CAS Number: 146137-76-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR