2'-Hydroxy-5'-(trifluoromethoxy)acetophenone 98%

Warning

H32003-06 H32003-03 H32003-MD
AAH32003-MDEA 56.25 USD
AAH32003-MD AAH32003-06 AAH32003-03
2'-Hydroxy-5'-(trifluoromethoxy)acetophenone 98%
2'-Hydroxy-5'-(trifluoromethoxy)acetophenone
Formula: C₉H₇F₃O₃
Melting Pt: 28…31 °C
Storage Temperature: Ambient
MDL Number: MFCD01091002
CAS Number: 146575-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR