6-Chloro-2-fluoro-3-methoxybenzonitrile 97%, JRD

Danger

H31973-06 H31973-03
AAH31973-06EA 239.76 USD
AAH31973-06 AAH31973-03
6-Chloro-2-fluoro-3-methoxybenzonitrile 97%, JRD
6-Chloro-2-fluoro-3-methoxybenzonitrile
Formula: C₈H₅ClFNO
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD09832269
CAS Number: 1017777-72-8
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR