3-Ethoxy-2,4-difluoroaniline 97%, JRD

Danger

H31965-03 H31965-06
AAH31965-03EA 94.86 USD
AAH31965-03 AAH31965-06
3-Ethoxy-2,4-difluoroaniline 97%, JRD
3-Ethoxy-2,4-difluoroaniline
Formula: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD09258726
CAS Number: 1017778-10-7
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR