3,4,5-Trifluorobenzenesulfonamide 97%

Warning

H31964-03 H31964-06
AAH31964-03EA 132.56 USD
AAH31964-03 AAH31964-06
3,4,5-Trifluorobenzenesulfonamide 97%
3,4,5-Trifluorobenzenesulphonamide
Formula: C₆H₄F₃NO₂S
MW: 211.16
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD09800668
CAS Number: 913472-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR