3-Chlorothiophene-2-boronic acid pinacol ester 95%
|
Warning
|
H31951-MD
AAH31951-MDEA
159.51
USD
AAH31951-MD
3-Chlorothiophene-2-boronic acid pinacol ester 95%
3-Chlorothiophene-2-boronic acid pinacol ester
Formula:
C₁₀H₁₄BClO₂S MW: 244.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09837624 CAS Number: 1040281-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...