2-Amino-4-(2,4-difluorophenyl)thiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H31948-04 H31948-ME
AAH31948-MEEA 57.47 USD
AAH31948-ME AAH31948-04
2-Amino-4-(2,4-difluorophenyl)thiazole 97%
2-Amino-4-(2,4-difluorophenyl)thiazole
Formula: C₉H₆F₂N₂S
MW: 212.22
Storage Temperature: Ambient
MDL Number: MFCD00472891
CAS Number: 105512-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR