2-Phenylquinoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

H31920-06 H31920-03
AAH31920-03EA 74.39 USD
AAH31920-03 AAH31920-06
2-Phenylquinoline 99+%
2-Phenylquinoline
Formula: C₁₅H₁₁N
MW: 205.26 g/mol
Boiling Pt: 363 °C
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00011568
CAS Number: 612-96-4
EINECS: 210-326-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR