6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Warning

H31910-MD H31910-03
AAH31910-MDEA 82.68 USD
AAH31910-MD AAH31910-03
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD01717328
CAS Number: 57196-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR