4-Fluoro-3-methoxybenzyl alcohol 95%

Warning

Synonyms: (4-Fluoro-3-methoxyphenyl)methanol

H31897-09 H31897-03
AAH31897-09EA 328.36 USD
AAH31897-09 AAH31897-03
4-Fluoro-3-methoxybenzyl alcohol 95%
4-Fluoro-3-methoxybenzyl alcohol
Formula: C₈H₉FO₂
MW: 156.16 g/mol
Melting Pt: 47…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00272133
CAS Number: 128495-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR