2-(Trifluoromethyl)-p-anisidine 97%, JRD

Danger

Synonyms: 4-Methoxy-2-(trifluoromethyl)aniline

H31872-MD H31872-03
AAH31872-03EA 197.58 USD
AAH31872-03 AAH31872-MD
2-(Trifluoromethyl)-p-anisidine 97%, JRD
2-(Trifluoromethyl)-p-anisidine
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972660
CAS Number: 53903-49-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR